Learn More
Alkali Scientific Aluminum Canisters for Blood Bag # 4R 9951
SDP
Catalog No. 5000663124
Encompass_Preferred
Canisters are constructed of light aluminum and are designed for most popular brands of of Storage Bags, or use our Universal line of Storage Canisters. All of Canisters have both a locking handle and thumb hole and can be autoclaved. Stainless Steel Canister Frames are available for all Canisters and offer a range of heights to maximize your freezer canister storage space. Description: BAXTER/FENWAL Blood Bag 50 mL Canister Blood Bag Number: 4R 9951 Dimensions L x D x H 6 7/8 x 3 9/16 x 7/16″ 163 x 90 x 11mm Frames Available F C 4R9951 5 F C 4R9951 7 F C 4R9951 8 Platform Dividers Available P20 4R9951 P24 4R9951 P31 4R9951 P39 4R9951
Catalog No. 50-006-63124 Supplier Alkali Scientific Supplier No. C4R9951
Canisters are constructed of light aluminum and are designed for most popular brands of of Storage Bags, or use our Universal line of Storage Canisters. All of Canisters have both a locking handle and thumb hole and can be autoclaved. Stainless Steel Canister Frames are available for all Canisters and offer a range of heights to maximize your freezer canister storage space. Description: BAXTER/FENWAL Blood Bag 50 mL Canister Blood Bag Number: 4R 9951 Dimensions L x D x H 6 7/8 x 3 9/16 x 7/16″ 163 x 90 x 11mm Frames Available F C 4R9951 5 F C 4R9951 7 F C 4R9951 8 Platform Dividers Available P20 4R9951 P24 4R9951 P31 4R9951 P39 4R9951