Learn More
Alkali Scientific Aluminum Canisters for Blood Bag # 4R 9959 & 4R 5464
SDP
Catalog No. 5000662125
Encompass_Preferred
Canisters are constructed of light aluminum and are designed for most popular brands of of Storage Bags, or use our Universal line of Storage Canisters. All of Canisters have both a locking handle and thumb hole and can be autoclaved. Stainless Steel Canister Frames are available for all Canisters and offer a range of heights to maximize your freezer canister storage space. Description: BAXTER/FENWAL Blood Bag 1000 mL Canister Blood Bag Number 4R 9959 4R 5464 Dimensions L x D x H 13 3/8 x 5 1/2 x 1/2″ 340 x 140 x 12mm Frames Available F C 4R9959 3 F C 4R9959 4 F C 4R9959 5 Platform Dividers Available P20 4R9959 P24 4R9959 P31 4R9959 P39 4R9959
Catalog No. 50-006-62125 Supplier Alkali Scientific Supplier No. C4R9959
Canisters are constructed of light aluminum and are designed for most popular brands of of Storage Bags, or use our Universal line of Storage Canisters. All of Canisters have both a locking handle and thumb hole and can be autoclaved. Stainless Steel Canister Frames are available for all Canisters and offer a range of heights to maximize your freezer canister storage space. Description: BAXTER/FENWAL Blood Bag 1000 mL Canister Blood Bag Number 4R 9959 4R 5464 Dimensions L x D x H 13 3/8 x 5 1/2 x 1/2″ 340 x 140 x 12mm Frames Available F C 4R9959 3 F C 4R9959 4 F C 4R9959 5 Platform Dividers Available P20 4R9959 P24 4R9959 P31 4R9959 P39 4R9959