Learn More
Alkali Scientific Aluminum Canisters for Blood Bag # 7400 2, 7403 2, 7410 2 & 7025 2
SDP
Catalog No. 5000662652
Encompass_Preferred
Canisters are constructed of light aluminum and are designed for most popular brands of of Storage Bags, or use our Universal line of Storage Canisters. All of Canisters have both a locking handle and thumb hole and can be autoclaved. Stainless Steel Canister Frames are available for all Canisters and offer a range of heights to maximize your freezer canister storage space. Description: CHARTERMED Blood Bag Canister Blood Bag Number 7400 2 7403 2 7410 2 7025 2 Dimensions L x D x H 9 x 9 x 7/16″ 229 x 229 x 11mm Frames Available F C 7400 2 Platform Dividers Available P20 7400 P24 7400 P31 7400 P39 7400
Catalog No. 50-006-62652 Supplier Alkali Scientific Supplier No. C7400
Canisters are constructed of light aluminum and are designed for most popular brands of of Storage Bags, or use our Universal line of Storage Canisters. All of Canisters have both a locking handle and thumb hole and can be autoclaved. Stainless Steel Canister Frames are available for all Canisters and offer a range of heights to maximize your freezer canister storage space. Description: CHARTERMED Blood Bag Canister Blood Bag Number 7400 2 7403 2 7410 2 7025 2 Dimensions L x D x H 9 x 9 x 7/16″ 229 x 229 x 11mm Frames Available F C 7400 2 Platform Dividers Available P20 7400 P24 7400 P31 7400 P39 7400