Learn More
Alkali Scientific Aluminum Canisters for Blood Bag # 791 05, 791 05U & 8 0346 0
SDP
Catalog No. 5000662654
Encompass_Preferred
Canisters and frames are constructed from light aluminum, providing strength, durability, and easy handling in laboratory and clinical environments. These canisters are designed to securely hold a variety of blood bag brands including Baxter Fenwal, Chartmed, Gambo, Stericon, Medsep, and Origen, ensuring compatibility across multiple collection and processing systems. The sturdy aluminum frame supports stable placement and safe handling during blood processing, storage, or transport. Available canister s include 50 mL, 250 mL, 500 mL, 750 mL, and 1000 mL to accommodate different volume requirements. Each canister features a locking handle mechanism that helps maintain a secure closure while handling or moving the unit. An integrated thumb hole provides additional grip and control, allowing users to safely lift and position the canister during laboratory procedures. The light construction reduces handling fatigue while maintaining structural integrity for repeated laboratory use.
Catalog No. 50-006-62654 Supplier Alkali Scientific Supplier No. CBBCV1
Canisters and frames are constructed from light aluminum, providing strength, durability, and easy handling in laboratory and clinical environments. These canisters are designed to securely hold a variety of blood bag brands including Baxter Fenwal, Chartmed, Gambo, Stericon, Medsep, and Origen, ensuring compatibility across multiple collection and processing systems. The sturdy aluminum frame supports stable placement and safe handling during blood processing, storage, or transport. Available canister s include 50 mL, 250 mL, 500 mL, 750 mL, and 1000 mL to accommodate different volume requirements. Each canister features a locking handle mechanism that helps maintain a secure closure while handling or moving the unit. An integrated thumb hole provides additional grip and control, allowing users to safely lift and position the canister during laboratory procedures. The light construction reduces handling fatigue while maintaining structural integrity for repeated laboratory use.