Learn More
Fine Science Tools ALUMINUM CASE- GREEN
SDP
Catalog No. NC2602311
Encompass
Colored Aluminum CasesDimensions:18 x 9 x 3cmAlloy / Material:AluminumAutoclave Safe:YesThis instrument case comes with a spiked silicone mat for secure storage of surgical instruments. Four colors available but may vary due to lot to lot variations. The case and silicone insert are autoclavable but should not be used for sterilization of surgical instruments.
Catalog No. NC2602311 Supplier Fine Science Tools Supplier No. 2033001
Colored Aluminum CasesDimensions:18 x 9 x 3cmAlloy / Material:AluminumAutoclave Safe:YesThis instrument case comes with a spiked silicone mat for secure storage of surgical instruments. Four colors available but may vary due to lot to lot variations. The case and silicone insert are autoclavable but should not be used for sterilization of surgical instruments.