Learn More
Fine Science Tools ALUMINUM CASE- YELLOW
SDP
Supplier:  Fine Science Tools 2033003
Encompass
Colored Aluminum CasesDimensions:18 x 9 x 3cmAlloy / Material:AluminumAutoclave Safe:YesThis instrument case comes with a spiked silicone mat for secure storage of surgical instruments. Four colors available but may vary due to lot to lot variations. The case and silicone insert are autoclavable but should not be used for sterilization of surgical instruments.
Catalog No. NC2602314