Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ALUMINUM CERIUM OXIDE NANOPOWDER ≤120»NM PARTICLE SIZE (BET) 99% TRACE METALS BASIS

Catalog No. 91924110G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-924-110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-924-110G Supplier Sigma Aldrich Supplier No. 91924110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Aluminum cerium oxide finds applications in in defense, automotive, aerospace and electrical industries.Product of Engi-Mat Co.

Specifications

CAS 12014-44-7
Molecular Formula AlCeO3
Linear Formula AlCeO3
MDL Number MFCD02687050
Quantity 10 g
Synonym Cerium aluminate; Cerium aluminum oxide
Molecular Weight (g/mol) 215.1
Percent Purity 99% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.