Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Chloride, 6 N, Ricca Chemical
SDP

Catalog No. 57016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
570-16 500 mL Natural Poly Bottle
570-4 120 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 570-16 Supplier Ricca Chemical Company Supplier No. 57016
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7784-13-6 , 7732-18-5 , 7440-44-0
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
Synonym Aluminum (III) Chloride Hexahydrate
PubChem CID 24564
ChEBI CHEBI:30115
IUPAC Name aluminium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Color Colorless to Green
CAS Min % 32.17, 66.0, 0.13
CAS Max % 33.48, 68.0, 0.14
Grade Laboratory
Packaging Natural Poly Bottle
pH 2.5 to 3.5
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 6Normal
Chemical Name or Material Aluminum Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.