Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Chloride Hexahydrate, 99%, Reagent Grade, Solstice

Catalog No. 60045449 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-449 500 g Poly Bottle
60-045-448 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-449 Supplier Solstice Supplier No. 237078500G

missing translation for 'validMainframeMsg'

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Color White
pH 2.5 to 3.5 at 20°C
Physical Form Crystalline Solid
Quantity 500 g
Packaging Poly Bottle
Appearance Crystals or Crystalline Chunk(s) or Chunk(s) or Solid
Vapor Pressure 1mmHg (100°C)
Density 2.4g/cm3
UN Number NONH for all modes of transport
IUPAC Name aluminium(3+) hexahydrate trichloride
Formula Weight 241.43g/mol
Percent Purity 99%
Grade ReagentPlus
Chemical Name or Material Aluminum Chloride Hexahydrate
Show More Show Less
Hazard Statement P305 + P351 + P338
Precautionary Statement H315-H319
EINECSNumber 231-208-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.