Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Chloride Hexahydrate (Crystalline/Certified), Fisher Chemical

Catalog No. A573500 Shop All Fisherbrand Products
Change view
Click to view available options
Glass Bottle; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
A573-500 Glass Bottle; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A573-500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Melting Point 181°C
Color White
CAS 7784-13-6
pH 2.0
Physical Form Solid
Quantity 500 g
Molecular Formula AlCl3H12O6
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
IUPAC Name aluminium(3+) hexahydrate trichloride
Molecular Weight (g/mol) 241.42
Formula Weight 241.43
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Density 2.39g/cm³
Chemical Name or Material Aluminum Chloride Hexahydrate
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.