Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Chloride, Hexahydrate, USP, 95-102%, Spectrum™ Chemical
SDP

Catalog No. 18600473 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-473 50 kg Fiber Drum
18-600-469 100 g Poly Bottle
18-600-470 500 g Poly Bottle
18-600-471 2.5 kg Poly Pail
18-600-472 12 kg Poly Pail
18-600-474 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-600-473 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AL22550KGBL
Only null left
Add to Cart
Add to Cart
Request a Quote

AL225, 7784-13-6, AlCl3·6H2O

Spectrum™ Chemical Aluminum Chloride, Hexahydrate, USP is mainly used in topical formulations to treat hyperhidrosis (excessive sweating). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
IUPAC Name aluminium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

CAS Max % 100%
Quantity 50 kg
Percent Purity 95 to 102%
Grade USP
Packaging Fiber Drum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.