Learn More
Perkin Elmer US LLC Aluminum Covers - Qty. 400
Catalog No. 502112121
Encompass_Preferred
The Aluminum Sample Pans and Covers are used with nonvolatile solid samples, such as polymers and pharmaceuticals. They are routinely used for applications such as polymer melting, crystallization, glass transitions of polymers, thermoplastics, and thermosets.
Catalog No. 50-211-2121 Supplier Perkin Elmer US LLC Supplier No. 02191073
The Aluminum Sample Pans and Covers are used with nonvolatile solid samples, such as polymers and pharmaceuticals. They are routinely used for applications such as polymer melting, crystallization, glass transitions of polymers, thermoplastics, and thermosets.