Learn More
MilliporeSigma™ Supelco™ Aluminum crimp seal (without septum)
Shop All MilliporeSigma Supelco ProductsDescription
- Used for sealing 100 mL serum bottles
- Fitted with gray stopper during analysis of biogas emission in lagoons.
Specifications
Specifications
| Color | Silver |
| Diameter (Metric) Outer | 20 mm |
| Material | Aluminum |
| No. per Pack | 100 |
| Style | Tear-away |
| Quantity | 100 Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.