Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Aluminum Crimp Seals with Prefitted Septa for 8mm Crimp Top Vials

Catalog No. 0640619W Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Clear Septa, Clear
Red, Blue
White, Silver
Septum:
Clear PTFE/Red Rubber
Clear Polyethylene
White Virgin PTFE
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum
06-406-19W Red, Blue Clear PTFE/Red Rubber
03-377-49 Clear Septa, Clear Clear Polyethylene
03-397-10 White, Silver White Virgin PTFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-406-19W Supplier Thermo Scientific™ Supplier No. C400898B

missing translation for 'validMainframeMsg'

Thermo Scientific™ National Aluminum Crimp Seals provide a secure, leak-resistant seal that you can count on.

Thermo Scientific™ National Aluminum Crimp Seals provide a secure, leak-resistant seal that you can count on.

  • Available in a variety of colors and septum types
  • Aluminum seals must be applied with a crimping tool
TRUSTED_SUSTAINABILITY

Specifications

Color (Seal) Blue
Description Clear PTFE/red rubber septa; Blue
Color Red, Blue
Material PTFE, Rubber
Septum Clear PTFE/Red Rubber
Closure Size 8 mm
For Use With (Equipment) 8 mm Crimp Top Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.