Learn More
Alkali Scientific Aluminum Cryo Canes, 5 Place, Cryo Canes with Platform
SDP
Catalog No. 5000662209
Encompass_Preferred
Aluminum cryo canes are designed for efficient organization and storage of cryogenic vials in laboratory freezers and liquid nitrogen storage systems. These canes are available in two configurations, with vial platforms and without vial platforms, allowing flexibility for different storage requirements. The design accommodates commonly used cryogenic vial s for sample preservation and biological storage. Each cryo cane can hold five cryogenic vials d 1.0 mL, 1.2 mL, 1.5 mL, 1.8 mL, or 2.0 mL when using the vial platform. When the platform is not used, the cane can accommodate vials d 1.0 mL, 1.2 mL, 1.5 mL, 1.8 mL, 2.0 mL, or 5.0 mL. The light aluminum construction ensures durability while allowing easy handling and efficient sample organization in high capacity cryogenic storage environments.
Catalog No. 50-006-62209 Supplier Alkali Scientific Supplier No. LFC5L
Aluminum cryo canes are designed for efficient organization and storage of cryogenic vials in laboratory freezers and liquid nitrogen storage systems. These canes are available in two configurations, with vial platforms and without vial platforms, allowing flexibility for different storage requirements. The design accommodates commonly used cryogenic vial s for sample preservation and biological storage. Each cryo cane can hold five cryogenic vials d 1.0 mL, 1.2 mL, 1.5 mL, 1.8 mL, or 2.0 mL when using the vial platform. When the platform is not used, the cane can accommodate vials d 1.0 mL, 1.2 mL, 1.5 mL, 1.8 mL, 2.0 mL, or 5.0 mL. The light aluminum construction ensures durability while allowing easy handling and efficient sample organization in high capacity cryogenic storage environments.