Learn More
Premier Lab Supply Straight Wall Aluminum Cups Producing 40mm Pellets – PN# AC-538
SDP
Supplier:  Premier Lab Supply AC538
Encompass_Preferred
Straight Wall Aluminum Cups with Dimensions 38.5mm x 9.5mm (PN# AC-538) – Suitable for producing 40mm pellets – PREMIER’s aluminum cups are made with a strong and durable aluminum alloy that provides stable backbone for pressed pellets. Thin wall material ensures minimal back pressure and eases reproducibility of pressed pellets. – 600 per bag.
Catalog No. NC0633158