Learn More
Electron Microscopy Sciences Aluminum Dish, Large
SDP
Catalog No. 5024499
Encompass_Preferred
Inexpensive, disposable aluminum foil dishes with crimped sides, flat smooth bottoms, and a finger tab handle on the rim. They may be used for routine weighing and moisture testing, as well as an embedding mold.
Size: Large
Catalog No. 50-244-99 Supplier Electron Microscopy Sciences Supplier No. 7005210
Inexpensive, disposable aluminum foil dishes with crimped sides, flat smooth bottoms, and a finger tab handle on the rim. They may be used for routine weighing and moisture testing, as well as an embedding mold.
Size: Large