Learn More
Electron Microscopy Sciences Aluminum Dish, Medium
SDP
Catalog No. 5024505
Encompass_Preferred
Inexpensive, disposable aluminum foil dishes with crimped sides, flat smooth bottoms, and a finger tab handle on the rim. They may be used for routine weighing and moisture testing, as well as an embedding mold.
Size: Medium
Catalog No. 50-245-05 Supplier Electron Microscopy Sciences Supplier No. 7005001
Inexpensive, disposable aluminum foil dishes with crimped sides, flat smooth bottoms, and a finger tab handle on the rim. They may be used for routine weighing and moisture testing, as well as an embedding mold.
Size: Medium