Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Aluminum-doped zinc oxide ink for spin coating/slot-die coating

Catalog No. 90109225ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-109-225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-109-225ML Supplier Sigma Aldrich Supplier No. 90109225ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Helios'Ink H-DZ41006 semiconductive ink developed for the printed electronics is particularly well suited for OPV and PV.This ink shows great performance on spin coating and slot die coating and is compatible with various flexible substrates (Kapton,(R) PET, PET/ITO, etc.) but also glass/ITO.The use of Helios'Ink H-DZ41006 semiconductive ink is greatly improving the devices performances by reducing the roughness of the TCE without a negative impact on the electrical conductivity and the transparency. Life time of the devices is also increased.The use of Helios'Ink H-DZ41006 provides ETL layer with high performances. In addition, compared to standard ETL, the PCE not retained even with higher thickness.Product of Genes'Ink.

Specifications

Linear Formula ZnO
Quantity 25 mL
Synonym AZO dispersion; AZO ink; Al-doped ZnO ink; HeliosInk H-DZ41006 semiconductive ink
Biological Activity Central nervous system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.