Learn More
Transene Inc Alumionum Etchant - Type A
SDP
Catalog No. NC1653938
Encompass_Preferred
Transene Aluminum Etchants are stable, non-toxic preparations used to etch aluminum metallizations on silicon devices and in integrated circuit applications. Aluminum contacts are defined, and interconnections are formed. These Aluminum Etchants, formulated with unique properties, easily overcome many of the difficulties experienced in aluminum etch processes.
Catalog No. NC1653938 Supplier Transene Inc Supplier No. ALUMINUMETCHAA
Transene Aluminum Etchants are stable, non-toxic preparations used to etch aluminum metallizations on silicon devices and in integrated circuit applications. Aluminum contacts are defined, and interconnections are formed. These Aluminum Etchants, formulated with unique properties, easily overcome many of the difficulties experienced in aluminum etch processes.