Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Etch (Keller's II), Reagents

Catalog No. 01335376
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-376 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-376 Supplier Reagents Holdings Llc Supplier No. CE230000100A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aluminum Etch (Keller's II), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Colorless to Slight Yellow
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.1374
Quantity 100 mL
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Aluminum Etch (Keller's II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.