Learn More
MSC Tanis Brushes Brush Holder, 36 in, Clear Anodized, 1 ea
Catalog No. 502818288
Encompass
This strip brush holder is designed for industrial applications, providing a robust solution for containing brushes. It features an aluminum backing with a clear finish and anodized coating, ensuring durability and corrosion resistance in various environments.
  • Constructed from aluminum with a clear finish anodized backing
  • Overall length of 36 in.
  • Features a 45 degree aluminum profile
  • Designed to accommodate #4 single Y track edging
  • Backing dimensions are 3/16 in. height and 3/16 in. width
Catalog No. 50-281-8288 Supplier MSC Supplier No. 09301615
This strip brush holder is designed for industrial applications, providing a robust solution for containing brushes. It features an aluminum backing with a clear finish and anodized coating, ensuring durability and corrosion resistance in various environments.
  • Constructed from aluminum with a clear finish anodized backing
  • Overall length of 36 in.
  • Features a 45 degree aluminum profile
  • Designed to accommodate #4 single Y track edging
  • Backing dimensions are 3/16 in. height and 3/16 in. width