Learn More
Cole-Parmer Heavy-Duty Aluminum Foil, 18" Wide; 500 Ft/Box
Supplier:  Cole-Parmer 0627505
Encompass_Preferred
All-purpose foil used for freezer stroage, wrapping, sealing, autoclaving, lining, and weighing or transfering samples. Packaged in a dispenser box with serrated cutting strip.
Catalog No. NC9802400