Learn More
Andwin Scientific ALUMINUM FOIL FOR 96W 100/CS
SDP
Catalog No. NC9933225
Encompass_Preferred
The Aluminum Foil is specially made to fit 96-well plates. It has one partial-width end tab with no perforation. The foil is an excellent product for use in PCR and cold storage applications. This product has a dimension of 12.7 x 7.8 cm (5 x 3") and
Catalog No. NC9933225 Supplier Andwin Scientific Supplier No. 60941126
The Aluminum Foil is specially made to fit 96-well plates. It has one partial-width end tab with no perforation. The foil is an excellent product for use in PCR and cold storage applications. This product has a dimension of 12.7 x 7.8 cm (5 x 3") and