Learn More
Electron Microscopy Sciences Aluminum Foil Squares, 4 x 4", 0.001" thick
SDP
Catalog No. NC1466903
Encompass_Preferred
Aluminum foil squares interleaved for easy separation. Weigh samples quickly and economically. Cap labware for storage or autoclaving.
Size: 4 x 4"
Thickness: 0.001"
Catalog No. NC1466903 Supplier Electron Microscopy Sciences Supplier No. 7005604
Aluminum foil squares interleaved for easy separation. Weigh samples quickly and economically. Cap labware for storage or autoclaving.
Size: 4 x 4"
Thickness: 0.001"