Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Aluminum Foil Tape

Catalog No. 19072120 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
60 yd. x 1 in.
60 yd. x 2 in.
Quantity:
24 Rolls/Cs.
36 Rolls/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Quantity
19-072-120 60 yd. x 1 in. 36 Rolls/Cs.
19-072-035 60 yd. x 2 in. 24 Rolls/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-072-120 Supplier 3M™ Supplier No. 7100053741
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Aluminum Foil Tape is ideal for numerous applications across a broad range of industries.

  • Thermally conductive tape helps dissipate heat and improves heating and cooling efficiency to protect temperature sensitive materials
  • Resistant to chemicals to protect surfaces during de-paint process and other chemical masking operations
  • Highly engineered acrylic adhesive provides long term durability, but removes cleanly after harsh masking applications
  • Comprised of a dead soft aluminum foil backing and coated with a highly engineered pressure sensitive acrylic adhesive
  • Ideal for heat shielding, heat reflecting, chemical masking, light enhancement, chemical milling, seaming, sealing and de-paint operations.
  • Withstands exposure to flame, weathering, moisture and UV
  • Heat and light reflective tape protects surfaces and enhances lighting efficiency
  • Dead soft aluminum foil backing conforms to curved and uneven surfaces
  • Recognized by UL to UL 746C and Classified by UL to UL 723, Class L for low flammability rating
  • Contains plastic core center
TRUSTED_SUSTAINABILITY

Specifications

Product Type Aluminum Foil Tape
For Use With (Application) Painting, Stripping
Adhesive Type Acrylic
Color Silver
Dimensions (L x W) 60 yd. x 1 in.
Material Acrylic
Model 425
Quantity 36 Rolls/Cs.
Thickness (English) 4.6 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.