Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Aluminum Foil Tape

Catalog No. 19070923 Shop All 3M Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-070-923 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-923 Supplier 3M™ Supplier No. 7100053743
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M Company Aluminum Foil Tape is thermally conductive and helps dissipate heat while improving heating and cooling efficiency to protect temperature sensitive materials

  • Resistant to chemicals to protect surfaces during de-paint process and other chemical masking operations
  • Highly engineered acrylic adhesive provides long term durability, but removes cleanly after harsh masking applications
  • Withstands exposure to flame, weathering, moisture and UV
  • Heat and light reflective tape protects surfaces and enhances lighting efficiency
  • Dead soft aluminum foil backing conforms to curved and uneven surfaces
  • Ideal for a broad range of industries including heat shielding, heat reflecting, chemical masking, light enhancement, chemical milling, seaming, sealing and de-paint operations.
  • Comprised of a dead soft aluminum foil backing and coated with a highly engineered pressure sensitive acrylic adhesive
  • 0.75 in x 60 yd 4.6 mil
  • 3M Catalog# 425

TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Product Type Foil Tape
For Use With (Application) Stripping
Model 425
Quantity 48 Pk.
Dimensions (L x W x H) 60 yd. x 0.75 in. x 4.6 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.