Learn More
TA Instruments Aluminum Foil for TGA Smart-Seal Pans Pack of 100, Hermetic Seal Tzero Pans, Part Number 901683.901, Secure Loading of Atmosphere Sensitive Samples, for Thermogravimetric Analyzer TGA
Catalog No. 5000580689 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Aluminum Foil for TGA Smart-Seal Pans Pack Of 100 includes 100 pieces of aluminum foil designed to hermetically seal Tzero pans P/N 901683.901. These foils allow secure loading of atmosphere-sensitive samples in a thermogravimetric analyzer (TGA).
Catalog No. 50-005-80689 Supplier TA Instruments Supplier No. 956970.901
Waters TA Instruments Aluminum Foil for TGA Smart-Seal Pans Pack Of 100 includes 100 pieces of aluminum foil designed to hermetically seal Tzero pans P/N 901683.901. These foils allow secure loading of atmosphere-sensitive samples in a thermogravimetric analyzer (TGA).