Learn More
Andwin Scientific ALUMINUM FOILS FOR PCR AND COL
SDP
Catalog No. NC0605847
Encompass_Preferred
The sterile Aluminum Foils for PCR and Cold Storage gives a safe and dependable sealing area for all PCR plates. The foil is 38 m thick and is free of RNase, DNase, and nuclease. It is soft and non-permeable but is equipped with durable, medical-gra
Catalog No. NC0605847 Supplier Andwin Scientific Supplier No. 60941076
The sterile Aluminum Foils for PCR and Cold Storage gives a safe and dependable sealing area for all PCR plates. The foil is 38 m thick and is free of RNase, DNase, and nuclease. It is soft and non-permeable but is equipped with durable, medical-gra