Learn More
Alkali Scientific Alkali Scientific™ Aluminum Freezer Racks for 2" Mini Boxes, Vertical Configuration, 14 Box Capacity
SDP
Catalog No. 503028849
Encompass_Preferred
For 2″ Mini Boxes The Aluminum Vertical Mini Racks with Locking Rod are expertly crafted storage solutions designed for efficient and secure organization of samples in laboratory freezers and cryogenic storage systems. Made from premium-grade aluminum, these racks are engineered to withstand the rigors of low-temperature environments, ensuring durability and performance. The vertical orientation maximizes freezer space, allowing for optimal organization and easy access to samples. Dimensions H x W x D: 31 x 3 3/8 x 3 1/4 inch (788 x 86 x 82mm)Compartment Size H x W x D: 2 3/16 x 3 1/16 x 3 1/16 (54.5 x 77 x 77) **Lock device is ordered separately.
Catalog No. 50-302-8849 Supplier Alkali Scientific Supplier No. MCFA142
For 2″ Mini Boxes The Aluminum Vertical Mini Racks with Locking Rod are expertly crafted storage solutions designed for efficient and secure organization of samples in laboratory freezers and cryogenic storage systems. Made from premium-grade aluminum, these racks are engineered to withstand the rigors of low-temperature environments, ensuring durability and performance. The vertical orientation maximizes freezer space, allowing for optimal organization and easy access to samples. Dimensions H x W x D: 31 x 3 3/8 x 3 1/4 inch (788 x 86 x 82mm)Compartment Size H x W x D: 2 3/16 x 3 1/16 x 3 1/16 (54.5 x 77 x 77) **Lock device is ordered separately.