Learn More
Alkali Scientific Aluminum Freezer Racks for 3 S
SDP
Supplier:  Alkali Scientific CFA43
Encompass_Preferred
The Aluminum Vertical Rack for Standard 3in Boxes is an indispensable storage accessory engineered for optimal use within laboratory chest freezers and liquid nitrogen freezers Constructed from premium-grade aluminum this rack is robust enough to endure the extreme conditions of low-temperature storage environments while providing efficient organization of standard 3in storage boxes Product Features Premium Aluminum Construction Crafted from high-grade aluminum this vertical rack boasts superior durability and resistance to the cold temperatures and moisture found in chest and liquid nitrogen freezers Its corrosion-resistant finish ensures longevity and reliability making it an ideal choice for rigorous laboratory use Designed for Standard 3in Boxes This rack is precisely dimensioned to hold standard 3in boxes ensuring a secure and stable fit The snug accommodation of boxes prevents accidental slips or falls thereby protecting valuable samples from potential damage Maximize Stora
Catalog No. 50-302-9167