Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Glycinate 97.0+%, TCI America™
SDP

Catalog No. A1200500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1200500G 500 g
A120025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1200500G Supplier TCI America Supplier No. A1200500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13682-92-3
Molecular Formula C6H15AlN3O6
Molecular Weight (g/mol) 252.183
MDL Number MFCD00053645
InChI Key VBASKOIIBZSXOZ-UHFFFAOYSA-N
Synonym Aminoacetic Acid Dihydroxyaluminum Salt, Dihydroxyaluminum Aminoacetate
PubChem CID 129894307
IUPAC Name aluminum;2-aminoacetic acid
SMILES C(C(=O)O)N.C(C(=O)O)N.C(C(=O)O)N.[Al]

Specifications

Color White
Quantity 500 g
Formula Weight 135.05
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Aluminum Glycinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.