Learn More
Chemglass Life Sciences Aluminum Heat Transfer Collar, Minum-Ware
SDP
Catalog No. 50872167
Encompass_Preferred
Aluminum Heat Transfer Collar, Minum-Ware. For use in conjunction with MW-114 aluminum heat transfer block. Collar assists in the heating of the larger 3.0 and 5.0ml MW-80 conical reaction vials. Effects are similar to pushing the vial deeper into a sand bath. The collar is 3/4in thick and 1 1/2in square with a 20mm hole, split to allow easy placement around vial.
Catalog No. 50-872-167 Supplier Chemglass Life Sciences Supplier No. MW11201
Aluminum Heat Transfer Collar, Minum-Ware. For use in conjunction with MW-114 aluminum heat transfer block. Collar assists in the heating of the larger 3.0 and 5.0ml MW-80 conical reaction vials. Effects are similar to pushing the vial deeper into a sand bath. The collar is 3/4in thick and 1 1/2in square with a 20mm hole, split to allow easy placement around vial.