Learn More
Sigma Aldrich Fine Chemicals Biosciences Aluminum heating/cooling b1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7434741EA
Encompass
Each Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures.
Catalog No. 50-290-0569