Learn More
Sigma Aldrich Fine Chemicals Biosciences Aluminum heating/cooling block Holds 96 x 0.2 mL tubes in center plus 10 on size (4 for 1.5ml tubes, 6 for 0.5ml tubes) | 1EA
Catalog No. NC2185916
Encompass
Aluminum heating/cooling block Holds 96 x 0.2 mL tubes in center plus 10 on size (4 for 1.5ml tubes, 6 for 0.5ml tubes) | 1EA
This item is not returnable. View return policy
Catalog No. NC2185916 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7402701EA
Aluminum heating/cooling block Holds 96 x 0.2 mL tubes in center plus 10 on size (4 for 1.5ml tubes, 6 for 0.5ml tubes) | 1EA