Learn More
Medchemexpress LLC Aluminum Hydroxide | 21645-51-2 | MFCD00003420 | 78.00 | 1 G
SDP
Catalog No. 5000365494
Encompass_Preferred
Aluminum Hydroxide is an orally active main form of aluminum used as an adjuvant. Research on aluminum hydroxide-based adjuvants includes its repository effect, pro-phagocytic effect, and activation of the pro-inflammatory NLRP3 pathway. It functions as an adjuvant to compensate for the low inherent immunogenicity of subunit vaccines.
  • Not recommended to be frozen when used as a vaccine adjuvant.
  • Oral administration increases lipid peroxidation and decreases SOD activity in the brain.
  • Can dissolve in distilled water to form a homogeneous suspension.
Catalog No. 50-003-65494 Supplier Medchemexpress LLC Supplier No. 1GHYB1521
Aluminum Hydroxide is an orally active main form of aluminum used as an adjuvant. Research on aluminum hydroxide-based adjuvants includes its repository effect, pro-phagocytic effect, and activation of the pro-inflammatory NLRP3 pathway. It functions as an adjuvant to compensate for the low inherent immunogenicity of subunit vaccines.
  • Not recommended to be frozen when used as a vaccine adjuvant.
  • Oral administration increases lipid peroxidation and decreases SOD activity in the brain.
  • Can dissolve in distilled water to form a homogeneous suspension.