Learn More
Medchemexpress LLC Aluminum hydroxide adjuvant | 10 ML
SDP
Catalog No. 5000373491
Encompass_Preferred
Aluminum hydroxide adjuvant is a vaccine adjuvant containing 9-11 mg/mL aluminum. It induces differentiation of macrophages into specialized antigen-presenting cells. This adjuvant provides strong adsorption for negatively charged antigens, offers a significant sustained-release effect, and has high safety, effectively inducing immune responses. It is suitable for various veterinary vaccines, including inactivated or gene-engineered subunit vaccines.
  • Induces specialized antigen-presenting macrophage differentiation.
  • Strong adsorption for negatively charged antigens.
  • Significant sustained-release effect.
  • High safety, effective immune response induction.
  • Suitable for veterinary inactivated/gene-engineered subunit vaccines.
  • Use instructions: filter antigen, shake adjuvant, mix 1:9, incubate 10 min.
  • Storage: 2-8°C, use within 3 months of opening.
  • Avoid high temp/pressure, freeze/thaw.
Catalog No. 50-003-73491 Supplier Medchemexpress LLC Supplier No. HY15320510ML
Aluminum hydroxide adjuvant is a vaccine adjuvant containing 9-11 mg/mL aluminum. It induces differentiation of macrophages into specialized antigen-presenting cells. This adjuvant provides strong adsorption for negatively charged antigens, offers a significant sustained-release effect, and has high safety, effectively inducing immune responses. It is suitable for various veterinary vaccines, including inactivated or gene-engineered subunit vaccines.
  • Induces specialized antigen-presenting macrophage differentiation.
  • Strong adsorption for negatively charged antigens.
  • Significant sustained-release effect.
  • High safety, effective immune response induction.
  • Suitable for veterinary inactivated/gene-engineered subunit vaccines.
  • Use instructions: filter antigen, shake adjuvant, mix 1:9, incubate 10 min.
  • Storage: 2-8°C, use within 3 months of opening.
  • Avoid high temp/pressure, freeze/thaw.