Learn More
Sigma Aldrich Fine Chemicals Biosciences Aluminum hydroxide reagent grade | Purity: 50.0-57.5% Al (as Al2O3) basis (EDTA titration) | Mol Wt: 78 | 21645-51-2 | MFCD00003420 | 25G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 23918625G
Encompass
Aluminum hydroxide reagent grade | Purity: 50.0-57.5% Al (as Al2O3) basis (EDTA titration) | Mol Wt: 78 | 21645-51-2 | MFCD00003420 | 25G
Catalog No. 50-188-3888