Learn More
Strem, An Ascensus Company CAS# 555-31-7. 250g. Aluminum i-propoxide, 98+%. MFCD00008870
SDP
Supplier:  Strem, An Ascensus Company 931345250G
Encompass
CAS# 555-31-7. 250g. Aluminum i-propoxide, 98+%. MFCD00008870. Molecular Weight: 204.25. Molecular Formula: Al[OCH(CH3)2]3. Color/form: white pwdr. Strem# 93-1345. http://www.strem.com/catalog/v/93-1345/
Catalog No. 50-901-14035