Learn More
Glas Col LLC Evaporator Accessory
SDP
Catalog No. NC1519058
Encompass_Preferred
Aluminum insert block, 24 position for up to 15/16mm vials, for use with 109A YH-1.
Catalog No. NC1519058 Supplier Glas Col LLC Supplier No. 109A YH2416
Aluminum insert block, 24 position for up to 15/16mm vials, for use with 109A YH-1.