Learn More
Aluminum iodide, 99.99+% (metals basis)
Description
Aluminum iodide is employed as a catalyst to break certain kinds of C-O and N-O bonds. It cleaves aryl ethers and deoxygenates epoxides It can also be used for the manufacture of iodinated organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 360°C |
| Melting Point | 191°C |
| Color | Brown-Purple to White |
| Physical Form | Lump |
| Quantity | 25 g |
| Odor | Pungent |
| Sensitivity | Light and moisture sensitive |
| Assay Percent Range | >99.99% (Metals basis) |
| Density | 3.98 g/cm3 |
| Solubility Information | Soluble in water,alcohol and ether. |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | Keep container dry. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | DOT Class: 8, Packaging Group: II |
| EINECSNumber | 232-054-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.