Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Magnesium gauze, alloy 5056, 30 mesh woven from 0.23mm (0.009in) dia wire

Catalog No. p-7051298
Change view
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40906FL 75 x 75 mm
AA40906GT 150 x 150 mm
AA40906HB 300 x 300 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40906FL Supplier Thermo Scientific Chemicals Supplier No. 040906FL
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminum Magnesium gauze, Alloy 5056
Name Note Woven from 0.23mm (0.009 in.) dia. wire
Avg. Mol. Wt. or Mol. Wt. Range Al:Mg; 95:5 wt%
Quantity 75 x 75 mm
Mesh Size 30 Mesh
Odor Odorless
Form Wire Cloth
MDL Number MFCD00214039

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.