Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Metal Shot, 99%, Lab Grade, Innovating Science™
SCI_ED

Catalog No. S000504 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000504 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000504 Supplier Aldon Corporation Supplier No. IS10051
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Aluminum
CAS 7249-90-5
Quantity 500 g
Form Granular
Molecular Weight (g/mol) 26.98 g/mol
Percent Purity > 99.5%
Grade Laboratory

This substance or mixture has not been classified as hazardous according to the Globally Harmonized System (GHS) of Classification and Labeling of Chemicals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.