Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Aluminum-Nickel Alloy

Catalog No. 72240500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-240-500G 500 g
72-240-100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 72-240-500G Supplier Sigma Aldrich Supplier No. 72240500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Aluminum-nickel alloy can be used:For the reduction of nitriles, nitro compounds and oximes to primary amines.For the conversion of carbonyl compounds to hydrocarbons.As a reducing agent to convert Ar-X derivatives to Ar-H compounds.For the synthesis of Raney nickel catalyst to be used as an alkylating agent.To synthesize non-pyrophoric Ni catalyst by alkali leaching.For the synthesis of alloyed skeletal Ni catalysts.Raney is a registered trademark of W.R. Grace and Co.-Conn.

Specifications

CAS 12635-27-7
Quantity 500 g
MDL Number MFCD00167392
Synonym Nickel-aluminum alloy; Raney(R)-Nickel alloy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.