Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Aluminum-Nickel Catalyst

Catalog No. 221651100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-165-1100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 22-165-1100G Supplier Sigma Aldrich Supplier No. 221651100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Raney-type alloy for the preparation of Raney™ Nickel catalyst. ,Raney is a registered trademark of W.R. Grace and Co.-Conn.

Specifications

CAS 762-48-1
Density 0.905 g/mL at 25°C
Molecular Formula C3H8BF3O
Linear Formula BF3 -+ CH3CH2CH2OH
MDL Number MFCD00167015
Quantity 100 g
Molecular Weight (g/mol) 127.9 g/mol
Biological Activity Central Nervous System, Kidney
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.