Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum nickel isopropoxide, 99.9% (m.b.), 10% w/v in IPA, packaged under Ar in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog No. AA42376AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42376AC 25 mL
AA42376AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42376AC Supplier Thermo Scientific Chemicals Supplier No. 42376AC

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70504-57-3
MDL Number MFCD00143593

Specifications

Boiling Point 120°C (0.6 mmHg)
Packaging ChemSeal™ bottles
Quantity 25 mL
UN Number UN1219
Sensitivity Moisture sensitive
Formula Weight 584.92
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Aluminum nickel isopropoxide
Hazard Category H225-H315-H317-H319-H335-H336-H351
Hazard Statement GHS H Statement
H225-H351-H315-H319-H317-H336
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
TSCA No
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.