Learn More
Electron Microscopy Sciences Conductive Aluminum/Nickel Tape
SDP
Catalog No. 5028582
Encompass_Preferred
Electrically conductive and with a clean background, the tape is ideal for scanning electron microscopy (SEM). Pressed thin Al. Adhesive power/25mm width: 840 gf. Capacity after 60 min: 0.1mm.
Conductive resistivity: 0.008ohm/sq. Inch
Thickness: 0.09 mm
Pad: 0.035 mm
Adhesive: 0.040mm
Catalog No. 50-285-82 Supplier Electron Microscopy Sciences Supplier No. 77814
Electrically conductive and with a clean background, the tape is ideal for scanning electron microscopy (SEM). Pressed thin Al. Adhesive power/25mm width: 840 gf. Capacity after 60 min: 0.1mm.
Conductive resistivity: 0.008ohm/sq. Inch
Thickness: 0.09 mm
Pad: 0.035 mm
Adhesive: 0.040mm