Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Aluminum oxide

Catalog No. 642991100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-299-1100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 64-299-1100ML Supplier Sigma Aldrich Supplier No. 642991100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Aluminium oxide (Al2O3) is a metal oxide based engineered nanosized material that is used in a variety of industrial applications. It can be prepared by the hydrolysis of the alkoxide of Al2O3 and calcinating the particles in the presence of stabilizing agents.Al2O3 based nanoparticle can be used for a wide range applications such as ceramics, aerospace, wastewater treatment, abrasives, paints, catalysts, and flooring materials.No dispersant

Specifications

CAS 1344-28-1
Quantity 100 mL
Molecular Formula Al2O3
Linear Formula Al2O3
MDL Number MFCD00003424
Molecular Weight (g/mol) 101.96
Density 1.06 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.