Learn More
Sigma Aldrich Fine Chemicals Biosciences Aluminum oxide activated b1KG
Catalog No. 502898243
Encompass
The product is basic activated aluminum oxide (Alumina Al2O3) having an activity grade Brockman I. The mechanism of formation of protective scales of Al2O3 on surfaces of binary M-Al (M fe Ni or Co) alloys at high temperatures has been studied. Activated aluminum oxides having different acidities and water concentrations (having Brockmann activity I-V) can easily detect various types of water (namely free water pore water and bound water) on the aluminum oxide present in soils.
Catalog No. 50-289-8243 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1994431KG
The product is basic activated aluminum oxide (Alumina Al2O3) having an activity grade Brockman I. The mechanism of formation of protective scales of Al2O3 on surfaces of binary M-Al (M fe Ni or Co) alloys at high temperatures has been studied. Activated aluminum oxides having different acidities and water concentrations (having Brockmann activity I-V) can easily detect various types of water (namely free water pore water and bound water) on the aluminum oxide present in soils.