Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide, activated, neutral, gamma-phase, 99.9% (metals basis)

Catalog No. AA4326636
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4326636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4326636 Supplier Thermo Scientific Chemicals Supplier No. 04326636

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Aluminum oxide is used as an adsorbent, desiccating agent and in the manufacture of dental cements and refractories. It is used in protein extraction as a component in the preparation of tissues and in bushings, spindles, lapping, polishing, refractory coating for investment casting shells, decorative non-skid tile and pressure blasting. It is also used in varnishes, for textile impregnation, in cosmetics and as an intermediate in pharmaceutical production. It is a source of aluminum in various reactions. It is used as a catalyst in a variety of reactions such as the Claus process and in the dehydration of alcohols to alkenes and as a grinding or blending agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.9% (Metals basis)
Merck Index 14,356
Solubility Information Insoluble in water.
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Percent Purity 99.9%
Odor Odorless
Density 3.965g/cm3
Assay (metals basis)
Chemical Name or Material Aluminum oxide, Activated, neutral, gamma-phase
Show More Show Less
EINECSNumber 215-691-6
RTECSNumber BD1200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.